Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08007447.hg.1 |
TC08000545.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08007447.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08007447.hg.1 |
ENST00000540020.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08007447.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000517696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000517696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000517696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000517696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000517696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000517696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000518673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000518673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800156144.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800156144.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800156144.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800156144.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800156144.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800156144.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800156144.hg.1 |
TC0800008179.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000220751.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000517696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000517696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000517696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000517696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000517696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000517696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000518673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000518673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
ENST00000522965.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008179.hg.1 |
NM_003821.5 |
Refseq |