Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000540673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000540673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000540673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000540673.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000544871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000544871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000544871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000544871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000544871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000615991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000615991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000615991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
ENST00000615991.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_001300941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_001300941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_001300941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_001300941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_001300941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200011081.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000311915.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000398055.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000540673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000540673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000540673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000540673.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000544871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000544871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000544871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000544871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000544871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000615991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000615991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000615991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
ENST00000615991.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_001300940.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_001300941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_001300941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_001300941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_001300941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_001300941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_152440.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200011081.hg.1 |
NM_152440.4 |
Refseq |