Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000452115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000452115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000452115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000452115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000452115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000452115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000452115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000478130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000478130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000478130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000478130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000478130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
ENST00000478130.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002994.hg.1 |
ENST00000263795.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002994.hg.1 |
ENST00000381246.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002994.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002994.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15002994.hg.1 |
TC15000337.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500150210.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150210.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150210.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500150210.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500150210.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150210.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150210.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500150210.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500150210.hg.1 |
TC1500007077.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000263795.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000381246.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000452115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000452115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000452115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000452115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000452115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000452115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000452115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000478130.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000478130.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000478130.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000478130.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000478130.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
ENST00000478130.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_001167941.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007077.hg.1 |
NM_024908.3 |
Refseq |