Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012422.hg.1 |
ENST00000357780.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012422.hg.1 |
ENST00000357780.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012422.hg.1 |
ENST00000357780.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012422.hg.1 |
ENST00000357780.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012422.hg.1 |
ENST00000357780.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012422.hg.1 |
ENST00000357780.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012422.hg.1 |
ENST00000357780.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012422.hg.1 |
ENST00000357780.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012422.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012422.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012422.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012422.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012422.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012422.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012422.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012422.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027809.hg.1 |
ENST00000357780.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027809.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027809.hg.1 |
TC11002327.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027807.hg.1 |
ENST00000357780.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027807.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027807.hg.1 |
TC11002327.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027806.hg.1 |
ENST00000357780.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027806.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027806.hg.1 |
TC11002327.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100195387.hg.1 |
ENST00000357780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195387.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195387.hg.1 |
ENST00000357780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195387.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195387.hg.1 |
TC1100012422.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100195388.hg.1 |
ENST00000357780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195388.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195388.hg.1 |
ENST00000357780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195388.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195388.hg.1 |
TC1100012422.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100012422.hg.1 |
ENST00000357780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012422.hg.1 |
ENST00000357780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012422.hg.1 |
ENST00000357780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012422.hg.1 |
ENST00000357780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012422.hg.1 |
ENST00000357780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012422.hg.1 |
ENST00000357780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012422.hg.1 |
ENST00000357780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012422.hg.1 |
ENST00000357780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012422.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012422.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012422.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012422.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012422.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012422.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012422.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012422.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195386.hg.1 |
ENST00000357780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195386.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195386.hg.1 |
ENST00000357780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195386.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195386.hg.1 |
TC1100012422.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100195390.hg.1 |
ENST00000357780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195390.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195390.hg.1 |
ENST00000357780.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195390.hg.1 |
NM_000482.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195390.hg.1 |
TC1100012422.hg.1 |
Clariom D TC |