Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013877.hg.1 |
TC08001083.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013877.hg.1 |
ENST00000341513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013877.hg.1 |
ENST00000413272.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08013877.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000418860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000418860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000418860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000418860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
ENST00000418860.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167739.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167739.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167739.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800167739.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800167739.hg.1 |
TC0800009967.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000413272.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000418860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000418860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000418860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000418860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
ENST00000418860.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009967.hg.1 |
NM_001010906.1 |
Refseq |