Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008968.hg.1 |
ENST00000403084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008968.hg.1 |
NM_001130861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22008968.hg.1 |
TC22000498.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
ENST00000403084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
ENST00000403084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
ENST00000403084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
ENST00000403084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
ENST00000403084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
ENST00000403084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
ENST00000403084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
ENST00000403084.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
ENST00000406028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
ENST00000406028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
ENST00000406028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
ENST00000406028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
ENST00000406028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
ENST00000406028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
ENST00000413119.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
ENST00000413119.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
ENST00000413119.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
ENST00000413119.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
ENST00000413119.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
ENST00000413119.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
ENST00000413119.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
ENST00000618236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
ENST00000618236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
ENST00000618236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
NM_001130861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
NM_001130861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
NM_001130861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
NM_001130861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
NM_001130861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
NM_001130861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
NM_001130861.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
NM_003277.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
NM_003277.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
NM_003277.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
NM_003277.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
NM_003277.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007987.hg.1 |
NM_003277.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200156983.hg.1 |
TC2200007987.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200156983.hg.1 |
ENST00000403084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156983.hg.1 |
NM_001130861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200156983.hg.1 |
ENST00000403084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200156983.hg.1 |
NM_001130861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
ENST00000403084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
ENST00000403084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
ENST00000403084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
ENST00000403084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
ENST00000403084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
ENST00000403084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
ENST00000403084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
ENST00000403084.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
ENST00000406028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
ENST00000406028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
ENST00000406028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
ENST00000406028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
ENST00000406028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
ENST00000406028.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
ENST00000413119.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
ENST00000413119.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
ENST00000413119.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
ENST00000413119.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
ENST00000413119.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
ENST00000413119.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
ENST00000413119.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
ENST00000618236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
ENST00000618236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
ENST00000618236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
NM_001130861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
NM_001130861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
NM_001130861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
NM_001130861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
NM_001130861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
NM_001130861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
NM_001130861.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
NM_003277.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
NM_003277.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
NM_003277.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
NM_003277.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
NM_003277.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007987.hg.1 |
NM_003277.3 |
Refseq |