Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014798.hg.1 |
TC03000827.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014798.hg.1 |
ENST00000356517.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03014798.hg.1 |
NM_207365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009189.hg.1 |
ENST00000356517.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009189.hg.1 |
ENST00000356517.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009189.hg.1 |
ENST00000356517.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009189.hg.1 |
ENST00000356517.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009189.hg.1 |
ENST00000356517.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009189.hg.1 |
ENST00000356517.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009189.hg.1 |
ENST00000356517.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009189.hg.1 |
ENST00000445270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009189.hg.1 |
ENST00000445270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009189.hg.1 |
ENST00000445270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009189.hg.1 |
ENST00000445270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009189.hg.1 |
ENST00000445270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009189.hg.1 |
ENST00000445270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009189.hg.1 |
ENST00000445270.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009189.hg.1 |
NM_207365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009189.hg.1 |
NM_207365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009189.hg.1 |
NM_207365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009189.hg.1 |
NM_207365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009189.hg.1 |
NM_207365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009189.hg.1 |
NM_207365.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300009189.hg.1 |
NM_207365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009189.hg.1 |
ENST00000356517.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009189.hg.1 |
ENST00000356517.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009189.hg.1 |
ENST00000356517.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009189.hg.1 |
ENST00000356517.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009189.hg.1 |
ENST00000356517.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009189.hg.1 |
ENST00000356517.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009189.hg.1 |
ENST00000356517.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009189.hg.1 |
ENST00000445270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009189.hg.1 |
ENST00000445270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009189.hg.1 |
ENST00000445270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009189.hg.1 |
ENST00000445270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009189.hg.1 |
ENST00000445270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009189.hg.1 |
ENST00000445270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009189.hg.1 |
ENST00000445270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300009189.hg.1 |
NM_207365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009189.hg.1 |
NM_207365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009189.hg.1 |
NM_207365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009189.hg.1 |
NM_207365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009189.hg.1 |
NM_207365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009189.hg.1 |
NM_207365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300009189.hg.1 |
NM_207365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300165957.hg.1 |
ENST00000356517.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300165957.hg.1 |
ENST00000445270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300165957.hg.1 |
NM_207365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300165957.hg.1 |
ENST00000356517.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300165957.hg.1 |
ENST00000445270.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300165957.hg.1 |
NM_207365.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300165957.hg.1 |
TC0300009189.hg.1 |
Clariom D TC |