Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
ENST00000315489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
ENST00000315489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
ENST00000315489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
ENST00000315489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
ENST00000315489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
ENST00000315489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
ENST00000315489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
ENST00000315489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
ENST00000315489.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
ENST00000382696.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
ENST00000382696.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
ENST00000382696.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
ENST00000382696.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
ENST00000382696.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
ENST00000382696.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
ENST00000382696.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
ENST00000591681.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
ENST00000591681.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
ENST00000591681.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
ENST00000591681.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
NM_182577.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
NM_182577.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
NM_182577.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
NM_182577.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
NM_182577.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
NM_182577.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
NM_182577.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
NM_182577.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900009087.hg.1 |
NM_182577.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013438.hg.1 |
ENST00000315489.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013438.hg.1 |
ENST00000382696.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013438.hg.1 |
NM_182577.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19013438.hg.1 |
TC19000986.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
ENST00000315489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
ENST00000315489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
ENST00000315489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
ENST00000315489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
ENST00000315489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
ENST00000315489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
ENST00000315489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
ENST00000315489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
ENST00000315489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
ENST00000382696.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
ENST00000382696.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
ENST00000382696.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
ENST00000382696.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
ENST00000382696.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
ENST00000382696.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
ENST00000382696.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
ENST00000591681.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
ENST00000591681.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
ENST00000591681.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
ENST00000591681.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
NM_182577.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
NM_182577.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
NM_182577.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
NM_182577.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
NM_182577.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
NM_182577.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
NM_182577.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
NM_182577.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900009087.hg.1 |
NM_182577.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172148.hg.1 |
ENST00000315489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172148.hg.1 |
ENST00000382696.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172148.hg.1 |
ENST00000591681.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172148.hg.1 |
NM_182577.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900172148.hg.1 |
TC1900009087.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900172148.hg.1 |
ENST00000315489.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172148.hg.1 |
ENST00000382696.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172148.hg.1 |
ENST00000591681.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900172148.hg.1 |
NM_182577.2 |
Refseq |