Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046752.hg.1 |
TC01003006.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046752.hg.1 |
ENST00000359785.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046752.hg.1 |
ENST00000420377.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046752.hg.1 |
ENST00000525799.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046752.hg.1 |
ENST00000528414.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046752.hg.1 |
ENST00000538253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046752.hg.1 |
NM_001193431.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046752.hg.1 |
NM_012411.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01046752.hg.1 |
NM_015967.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214408.hg.1 |
ENST00000359785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214408.hg.1 |
ENST00000420377.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214408.hg.1 |
ENST00000525799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214408.hg.1 |
ENST00000528414.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214408.hg.1 |
ENST00000532224.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214408.hg.1 |
ENST00000538253.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214408.hg.1 |
NM_001193431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214408.hg.1 |
NM_001308297.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214408.hg.1 |
NM_012411.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214408.hg.1 |
NM_015967.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214408.hg.1 |
ENST00000359785.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214408.hg.1 |
ENST00000420377.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214408.hg.1 |
ENST00000525799.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214408.hg.1 |
ENST00000528414.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214408.hg.1 |
ENST00000532224.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214408.hg.1 |
ENST00000538253.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214408.hg.1 |
NM_001193431.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214408.hg.1 |
NM_001308297.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214408.hg.1 |
NM_012411.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214408.hg.1 |
NM_015967.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214408.hg.1 |
TC0100015333.hg.1 |
Clariom D TC |