Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006488.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006488.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006488.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006488.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006488.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006488.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006488.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006488.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006488.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006488.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006488.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006488.hg.1 |
NM_001011719.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006488.hg.1 |
NM_001011719.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006488.hg.1 |
NM_001011719.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006488.hg.1 |
NM_001011719.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006488.hg.1 |
NM_001011719.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006488.hg.1 |
NM_001011719.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006488.hg.1 |
NM_001011719.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006488.hg.1 |
NM_001011719.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006488.hg.1 |
NM_001011719.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006488.hg.1 |
NM_001011719.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000304.hg.1 |
TC0X000019.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000304.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000304.hg.1 |
NM_001011719.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006488.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006488.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006488.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006488.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006488.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006488.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006488.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006488.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006488.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006488.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006488.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006488.hg.1 |
NM_001011719.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006488.hg.1 |
NM_001011719.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006488.hg.1 |
NM_001011719.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006488.hg.1 |
NM_001011719.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006488.hg.1 |
NM_001011719.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006488.hg.1 |
NM_001011719.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006488.hg.1 |
NM_001011719.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006488.hg.1 |
NM_001011719.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006488.hg.1 |
NM_001011719.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006488.hg.1 |
NM_001011719.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00144538.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00144538.hg.1 |
NM_001011719.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00144538.hg.1 |
ENST00000381130.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00144538.hg.1 |
NM_001011719.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00144538.hg.1 |
TC0X00006488.hg.1 |
Clariom D TC |