Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIS18BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005267833.4 | XP_005267890.1 | 2 - 3 | 408 | 144 | |
XM_017021425.1 | XP_016876914.1 | 3 - 4 | 389 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |