Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000556455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000556455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000556455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000556455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000556455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000557603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000557603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000557603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000557603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000563329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000563329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000563329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000563329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000563329.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000565094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000565094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000565094.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000566478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000566478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
ENST00000566478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014616.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014616.hg.1 |
ENST00000553558.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014616.hg.1 |
ENST00000560393.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014616.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014616.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14014616.hg.1 |
TC14001290.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000556455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000556455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000556455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000556455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000556455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000557603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000557603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000557603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000557603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000563329.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000563329.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000563329.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000563329.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000563329.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000565094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000565094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000565094.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000566478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000566478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
ENST00000566478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400009655.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400171033.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400171033.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400171033.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400171033.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400171033.hg.1 |
ENST00000334988.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400171033.hg.1 |
ENST00000553558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400171033.hg.1 |
NM_001220484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400171033.hg.1 |
NM_203309.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400171033.hg.1 |
TC1400009655.hg.1 |
Clariom D TC |