Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006551.hg.1 |
TC0X000493.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006551.hg.1 |
ENST00000372691.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006551.hg.1 |
ENST00000372695.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006551.hg.1 |
NM_001080425.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006551.hg.1 |
NM_001127688.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007999.hg.1 |
ENST00000372691.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007999.hg.1 |
ENST00000372691.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007999.hg.1 |
ENST00000372691.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007999.hg.1 |
ENST00000372695.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007999.hg.1 |
ENST00000372695.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007999.hg.1 |
ENST00000372695.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007999.hg.1 |
ENST00000372695.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007999.hg.1 |
ENST00000372695.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007999.hg.1 |
ENST00000372695.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007999.hg.1 |
ENST00000372695.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007999.hg.1 |
NM_001080425.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007999.hg.1 |
NM_001080425.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007999.hg.1 |
NM_001080425.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007999.hg.1 |
NM_001080425.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007999.hg.1 |
NM_001080425.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007999.hg.1 |
NM_001080425.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007999.hg.1 |
NM_001127688.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007999.hg.1 |
NM_001127688.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007999.hg.1 |
NM_001127688.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007999.hg.1 |
NM_001127688.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007999.hg.1 |
NM_001127688.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006550.hg.1 |
TC0X000493.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006550.hg.1 |
ENST00000372695.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006550.hg.1 |
NM_001080425.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006550.hg.1 |
NM_001127688.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006554.hg.1 |
TC0X000493.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006554.hg.1 |
ENST00000372691.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006554.hg.1 |
ENST00000372695.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006554.hg.1 |
NM_001080425.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006554.hg.1 |
NM_001127688.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154545.hg.1 |
ENST00000372695.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154545.hg.1 |
NM_001080425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154545.hg.1 |
NM_001127688.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154545.hg.1 |
ENST00000372695.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154545.hg.1 |
NM_001080425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154545.hg.1 |
NM_001127688.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154545.hg.1 |
TC0X00007999.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00154549.hg.1 |
ENST00000372691.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154549.hg.1 |
ENST00000372695.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154549.hg.1 |
NM_001080425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154549.hg.1 |
NM_001127688.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154549.hg.1 |
ENST00000372691.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154549.hg.1 |
ENST00000372695.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154549.hg.1 |
NM_001080425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154549.hg.1 |
NM_001127688.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154549.hg.1 |
TC0X00007999.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00007999.hg.1 |
ENST00000372691.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007999.hg.1 |
ENST00000372691.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007999.hg.1 |
ENST00000372691.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007999.hg.1 |
ENST00000372695.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007999.hg.1 |
ENST00000372695.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007999.hg.1 |
ENST00000372695.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007999.hg.1 |
ENST00000372695.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007999.hg.1 |
ENST00000372695.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007999.hg.1 |
ENST00000372695.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007999.hg.1 |
ENST00000372695.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007999.hg.1 |
NM_001080425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007999.hg.1 |
NM_001080425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007999.hg.1 |
NM_001080425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007999.hg.1 |
NM_001080425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007999.hg.1 |
NM_001080425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007999.hg.1 |
NM_001080425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007999.hg.1 |
NM_001127688.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007999.hg.1 |
NM_001127688.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007999.hg.1 |
NM_001127688.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007999.hg.1 |
NM_001127688.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007999.hg.1 |
NM_001127688.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154546.hg.1 |
ENST00000372691.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154546.hg.1 |
ENST00000372695.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154546.hg.1 |
NM_001080425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154546.hg.1 |
NM_001127688.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154546.hg.1 |
ENST00000372691.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154546.hg.1 |
ENST00000372695.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00154546.hg.1 |
NM_001080425.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154546.hg.1 |
NM_001127688.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00154546.hg.1 |
TC0X00007999.hg.1 |
Clariom D TC |