Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024452.2 | NP_001019623.2 | 5 - 6 | 1003 | 176 | |
NM_001172285.1 | NP_001165756.1 | 5 - 6 | 1003 | 176 | ||
GenBank mRNA | AK309010.1 | - | 5 - 6 | 1003 | 176 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |