Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011527.hg.1 |
ENST00000402034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011527.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011527.hg.1 |
TC22000656.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
ENST00000437871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
ENST00000437871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
ENST00000437871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
ENST00000437871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
ENST00000437871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
ENST00000437871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
ENST00000437871.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
ENST00000437871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
ENST00000437871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
ENST00000437871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
ENST00000437871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
ENST00000437871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
ENST00000437871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
ENST00000437871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008445.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160667.hg.1 |
TC2200008445.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200160667.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160667.hg.1 |
ENST00000437871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160667.hg.1 |
NM_001193336.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160667.hg.1 |
ENST00000402034.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160667.hg.1 |
ENST00000437871.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160667.hg.1 |
NM_001193336.3 |
Refseq |