Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014765.hg.1 |
ENST00000300069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014765.hg.1 |
ENST00000560606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014765.hg.1 |
NM_194272.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014765.hg.1 |
TC15001553.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
ENST00000300069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
ENST00000300069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
ENST00000300069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
ENST00000300069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
ENST00000300069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
ENST00000300069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
ENST00000300069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
ENST00000300069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
ENST00000300069.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
ENST00000560606.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
ENST00000560606.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
ENST00000560606.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
ENST00000560606.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
ENST00000560606.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
ENST00000560606.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
ENST00000560606.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
ENST00000560606.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
NM_194272.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
NM_194272.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
NM_194272.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
NM_194272.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
NM_194272.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
NM_194272.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
NM_194272.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
NM_194272.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009756.hg.1 |
NM_194272.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
ENST00000300069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
ENST00000300069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
ENST00000300069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
ENST00000300069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
ENST00000300069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
ENST00000300069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
ENST00000300069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
ENST00000300069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
ENST00000300069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
ENST00000560606.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
ENST00000560606.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
ENST00000560606.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
ENST00000560606.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
ENST00000560606.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
ENST00000560606.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
ENST00000560606.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
ENST00000560606.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
NM_194272.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
NM_194272.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
NM_194272.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
NM_194272.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
NM_194272.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
NM_194272.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
NM_194272.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
NM_194272.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009756.hg.1 |
NM_194272.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500172072.hg.1 |
ENST00000300069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500172072.hg.1 |
ENST00000560606.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500172072.hg.1 |
NM_194272.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500172072.hg.1 |
ENST00000300069.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500172072.hg.1 |
ENST00000560606.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500172072.hg.1 |
NM_194272.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500172072.hg.1 |
TC1500009756.hg.1 |
Clariom D TC |