Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003122.hg.1 |
TC04000212.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003122.hg.1 |
ENST00000381980.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003122.hg.1 |
NM_001104629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003124.hg.1 |
TC04000212.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003124.hg.1 |
ENST00000381980.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003124.hg.1 |
ENST00000508175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003124.hg.1 |
NM_001104629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
ENST00000284437.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
ENST00000284437.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
ENST00000284437.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
ENST00000284437.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
ENST00000381980.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
ENST00000381980.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
ENST00000381980.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
ENST00000381980.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
ENST00000381980.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
ENST00000381980.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
ENST00000508175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
ENST00000508175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
ENST00000508175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
ENST00000508175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
NM_001104629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
NM_001104629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
NM_001104629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
NM_001104629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
NM_001104629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
NM_001104629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
NM_001104629.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
NM_018302.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
NM_018302.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
NM_018302.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
NM_018302.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007232.hg.1 |
NM_018302.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003125.hg.1 |
TC04000212.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003125.hg.1 |
ENST00000381980.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003125.hg.1 |
ENST00000508175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04003125.hg.1 |
NM_001104629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400149435.hg.1 |
ENST00000381980.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149435.hg.1 |
ENST00000508175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149435.hg.1 |
NM_001104629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400149435.hg.1 |
ENST00000381980.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149435.hg.1 |
ENST00000508175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149435.hg.1 |
NM_001104629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400149435.hg.1 |
TC0400007232.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400149432.hg.1 |
ENST00000381980.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149432.hg.1 |
NM_001104629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400149432.hg.1 |
ENST00000381980.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149432.hg.1 |
NM_001104629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400149432.hg.1 |
TC0400007232.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400149434.hg.1 |
ENST00000381980.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149434.hg.1 |
ENST00000508175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149434.hg.1 |
NM_001104629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400149434.hg.1 |
ENST00000381980.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149434.hg.1 |
ENST00000508175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400149434.hg.1 |
NM_001104629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400149434.hg.1 |
TC0400007232.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
ENST00000284437.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
ENST00000284437.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
ENST00000284437.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
ENST00000284437.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
ENST00000381980.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
ENST00000381980.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
ENST00000381980.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
ENST00000381980.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
ENST00000381980.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
ENST00000381980.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
ENST00000508175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
ENST00000508175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
ENST00000508175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
ENST00000508175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
NM_001104629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
NM_001104629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
NM_001104629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
NM_001104629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
NM_001104629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
NM_001104629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
NM_001104629.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
NM_018302.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
NM_018302.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
NM_018302.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
NM_018302.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007232.hg.1 |
NM_018302.2 |
Refseq |