Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035006.2 | NP_001030178.1 | 1 - 2 | 108 | 135 | |
GenBank mRNA | BI086293.1 | - | 1 - 2 | 79 | 135 | |
CD386294.1 | - | 1 - 2 | 127 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Inventoried Probe spans exons |