Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
aldo-keto reductase family 1, member C2 |
AKR1C2 |
AKR1C-pseudo, BABP, DD, DD-2, DD/BABP, DD2, DDH2, HAKRD, HBAB, MCDR2, SRXY8, TDD |
Anti-sense | Chr. 10 - 4987775 - 5018033 on NCBI Build GRCh38 | |
Downstream |
aldo-keto reductase family 1, member C3 |
AKR1C3 |
DD3, DDX, HA1753, HAKRB, HAKRe, HSD17B5, PGFS, hluPGFS |
56.3kb | Sense | Chr. 10 - 5048766 - 5107686 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |