Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005880.hg.1 |
ENST00000249842.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005880.hg.1 |
ENST00000395118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005880.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005880.hg.1 |
NM_201526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005880.hg.1 |
TC15000678.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
ENST00000249842.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
ENST00000249842.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
ENST00000249842.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
ENST00000249842.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
ENST00000249842.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
ENST00000249842.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
ENST00000249842.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
ENST00000395118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
ENST00000395118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
ENST00000395118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
ENST00000395118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
ENST00000395118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
ENST00000395118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
ENST00000559510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
ENST00000559510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
ENST00000560862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
ENST00000560862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
ENST00000560862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
NM_201526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
NM_201526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
NM_201526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
NM_201526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007839.hg.1 |
NM_201526.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005875.hg.1 |
ENST00000249842.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005875.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005875.hg.1 |
TC15000678.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005874.hg.1 |
ENST00000249842.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005874.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005874.hg.1 |
TC15000678.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500155779.hg.1 |
ENST00000249842.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155779.hg.1 |
ENST00000395118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155779.hg.1 |
ENST00000559510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155779.hg.1 |
ENST00000560862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155779.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500155779.hg.1 |
NM_201526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500155779.hg.1 |
ENST00000249842.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155779.hg.1 |
ENST00000395118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155779.hg.1 |
ENST00000559510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155779.hg.1 |
ENST00000560862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155779.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500155779.hg.1 |
NM_201526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500155779.hg.1 |
TC1500007839.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500155781.hg.1 |
ENST00000249842.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155781.hg.1 |
ENST00000395118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155781.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500155781.hg.1 |
NM_201526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500155781.hg.1 |
ENST00000249842.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155781.hg.1 |
ENST00000395118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155781.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500155781.hg.1 |
NM_201526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500155781.hg.1 |
TC1500007839.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500155772.hg.1 |
ENST00000249842.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155772.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500155772.hg.1 |
ENST00000249842.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155772.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500155772.hg.1 |
TC1500007839.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500155780.hg.1 |
ENST00000249842.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155780.hg.1 |
ENST00000395118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155780.hg.1 |
ENST00000560862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155780.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500155780.hg.1 |
NM_201526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500155780.hg.1 |
ENST00000249842.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155780.hg.1 |
ENST00000395118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155780.hg.1 |
ENST00000560862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155780.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500155780.hg.1 |
NM_201526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500155780.hg.1 |
TC1500007839.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
ENST00000249842.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
ENST00000249842.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
ENST00000249842.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
ENST00000249842.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
ENST00000249842.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
ENST00000249842.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
ENST00000249842.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
ENST00000395118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
ENST00000395118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
ENST00000395118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
ENST00000395118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
ENST00000395118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
ENST00000395118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
ENST00000559510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
ENST00000559510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
ENST00000560862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
ENST00000560862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
ENST00000560862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
NM_201526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
NM_201526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
NM_201526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
NM_201526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007839.hg.1 |
NM_201526.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500155771.hg.1 |
ENST00000249842.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155771.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500155771.hg.1 |
ENST00000249842.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500155771.hg.1 |
NM_005545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500155771.hg.1 |
TC1500007839.hg.1 |
Clariom D TC |