Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
ENST00000367175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
ENST00000367175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
ENST00000367175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
ENST00000367175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
ENST00000367175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
ENST00000367176.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
ENST00000367176.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
ENST00000367176.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
ENST00000367176.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
ENST00000367176.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
ENST00000367176.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
ENST00000367177.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
ENST00000367177.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
ENST00000367177.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
ENST00000367177.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
ENST00000367177.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
ENST00000367177.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
ENST00000367177.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
ENST00000496057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
ENST00000496057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
ENST00000496057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
ENST00000496057.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
NM_006338.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
NM_006338.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
NM_006338.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
NM_006338.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
NM_006338.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
NM_006338.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
NM_201630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
NM_201630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
NM_201630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
NM_201630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
NM_201630.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017043.hg.1 |
NM_201630.1 |
Refseq |