Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
8015207 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8015209 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8019575 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8015217 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8015216 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8015213 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8019577 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8015212 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8015208 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8019574 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8015206 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8015214 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8015210 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8019576 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
234691_PM_at |
- |
- |
Affymetrix Human Genome U219 Array |
11738938_at |
- |
- |
Affymetrix Human Genome U219 Array |
11738938_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019687.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019687.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019687.hg.1 |
TC17001483.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019689.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019689.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019689.hg.1 |
TC17001483.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019691.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019691.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019691.hg.1 |
TC17001483.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010652.hg.1 |
ENST00000398477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010652.hg.1 |
ENST00000398477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010652.hg.1 |
ENST00000398477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010652.hg.1 |
ENST00000398477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010652.hg.1 |
ENST00000398477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010652.hg.1 |
ENST00000542910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010652.hg.1 |
ENST00000542910.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010652.hg.1 |
NM_033032.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010652.hg.1 |
NM_033032.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010652.hg.1 |
NM_033032.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010652.hg.1 |
NM_033032.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010652.hg.1 |
NM_033032.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019690.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019690.hg.1 |
ENST00000542137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019690.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019690.hg.1 |
TC17001483.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019688.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019688.hg.1 |
ENST00000542137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019688.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17019688.hg.1 |
TC17001483.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010651.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010651.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010651.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010651.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010651.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010651.hg.1 |
ENST00000542137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010651.hg.1 |
ENST00000542137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010651.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010651.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010651.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010651.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010651.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184187.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184187.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184187.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184187.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184187.hg.1 |
TC1700010651.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700184192.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184192.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184192.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184192.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184192.hg.1 |
TC1700010651.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700184188.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184188.hg.1 |
ENST00000542137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184188.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184188.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184188.hg.1 |
ENST00000542137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184188.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184188.hg.1 |
TC1700010651.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700010651.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010651.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010651.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010651.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010651.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010651.hg.1 |
ENST00000542137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010651.hg.1 |
ENST00000542137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010651.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010651.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010651.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010651.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010651.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184190.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184190.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184190.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184190.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184190.hg.1 |
TC1700010651.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700184191.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184191.hg.1 |
ENST00000542137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184191.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184191.hg.1 |
ENST00000391419.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184191.hg.1 |
ENST00000542137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700184191.hg.1 |
NM_001123387.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700184191.hg.1 |
TC1700010651.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700010652.hg.1 |
ENST00000398477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010652.hg.1 |
ENST00000398477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010652.hg.1 |
ENST00000398477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010652.hg.1 |
ENST00000398477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010652.hg.1 |
ENST00000398477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010652.hg.1 |
ENST00000542910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010652.hg.1 |
ENST00000542910.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010652.hg.1 |
NM_033032.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010652.hg.1 |
NM_033032.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010652.hg.1 |
NM_033032.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010652.hg.1 |
NM_033032.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010652.hg.1 |
NM_033032.2 |
Refseq |
Human Genome U133 Plus 2.0 Array |
234691_PM_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
234691_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234691_at10 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234691_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234691_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234691_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234691_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234691_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234691_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234691_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234691_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234691_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
234691_at3 |
- |
- |
Human Genome U133 Set |
234691_PM_at |
- |
- |
Human Genome U133 Set |
234691_at |
- |
- |