Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
ENST00000537478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
ENST00000537478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
ENST00000537478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
ENST00000537478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
ENST00000537478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
ENST00000540814.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
ENST00000540814.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
ENST00000540814.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
ENST00000540814.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
ENST00000541065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
ENST00000541065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
ENST00000541065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
ENST00000541065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
ENST00000541065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
ENST00000541769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
ENST00000541769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
ENST00000541769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
ENST00000541769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
ENST00000541769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
ENST00000545853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
ENST00000545853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
ENST00000545853.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
NR_110034.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
NR_110034.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
NR_110034.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
NR_110034.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
NR_110034.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
NR_110035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
NR_110035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
NR_110035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
NR_110035.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
NR_110036.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
NR_110036.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
NR_110036.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
NR_110036.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
NR_110036.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012368.hg.1 |
NR_110036.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196823.hg.1 |
ENST00000537478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196823.hg.1 |
ENST00000540814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196823.hg.1 |
ENST00000541065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196823.hg.1 |
ENST00000541769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196823.hg.1 |
ENST00000545853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196823.hg.1 |
NR_110034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196823.hg.1 |
NR_110035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196823.hg.1 |
NR_110036.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196823.hg.1 |
ENST00000537478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196823.hg.1 |
ENST00000540814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196823.hg.1 |
ENST00000541065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196823.hg.1 |
ENST00000541769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196823.hg.1 |
ENST00000545853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196823.hg.1 |
NR_110034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196823.hg.1 |
NR_110035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196823.hg.1 |
NR_110036.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196823.hg.1 |
TC1200012368.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200196824.hg.1 |
ENST00000537478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196824.hg.1 |
ENST00000540814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196824.hg.1 |
ENST00000541065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196824.hg.1 |
ENST00000541769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196824.hg.1 |
ENST00000545853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196824.hg.1 |
NR_110034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196824.hg.1 |
NR_110035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196824.hg.1 |
NR_110036.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196824.hg.1 |
ENST00000537478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196824.hg.1 |
ENST00000540814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196824.hg.1 |
ENST00000541065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196824.hg.1 |
ENST00000541769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196824.hg.1 |
ENST00000545853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196824.hg.1 |
NR_110034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196824.hg.1 |
NR_110035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196824.hg.1 |
NR_110036.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196824.hg.1 |
TC1200012368.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
ENST00000537478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
ENST00000537478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
ENST00000537478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
ENST00000537478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
ENST00000537478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
ENST00000540814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
ENST00000540814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
ENST00000540814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
ENST00000540814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
ENST00000541065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
ENST00000541065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
ENST00000541065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
ENST00000541065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
ENST00000541065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
ENST00000541769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
ENST00000541769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
ENST00000541769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
ENST00000541769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
ENST00000541769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
ENST00000545853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
ENST00000545853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
ENST00000545853.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
NR_110034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
NR_110034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
NR_110034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
NR_110034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
NR_110034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
NR_110035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
NR_110035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
NR_110035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
NR_110035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
NR_110036.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
NR_110036.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
NR_110036.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
NR_110036.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
NR_110036.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012368.hg.1 |
NR_110036.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196827.hg.1 |
ENST00000540814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196827.hg.1 |
ENST00000541065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196827.hg.1 |
ENST00000541769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196827.hg.1 |
NR_110034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196827.hg.1 |
NR_110035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196827.hg.1 |
NR_110036.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196827.hg.1 |
ENST00000540814.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196827.hg.1 |
ENST00000541065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196827.hg.1 |
ENST00000541769.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200196827.hg.1 |
NR_110034.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196827.hg.1 |
NR_110035.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196827.hg.1 |
NR_110036.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200196827.hg.1 |
TC1200012368.hg.1 |
Clariom D TC |