Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FITM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203402.2 | NP_981947.1 | 2 | 854 | 60 | |
GenBank mRNA | BC139911.1 | - | 2 | 854 | 40147642 | 60 |
BI114004.1 | - | 2 | 667 | 60 | ||
BQ574746.1 | - | 1 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09170361 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29627770 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09170363 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09170372 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09170374 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29627775 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29627765 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29627762 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09170373 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09170358 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09170369 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29627779 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29627782 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P29627773 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P99625 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P99625 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 14146 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 14699 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P33487827 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P99625 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P99625 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09170369 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09170365 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09170363 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29627770 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09170374 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29627773 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29627762 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09170373 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29627782 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29627779 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09170366 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09170361 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09170358 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09170372 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29627775 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09170372 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29627773 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09170366 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09170363 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29627779 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09170369 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29627770 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09170361 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09170373 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29627762 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09170358 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09170374 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09170365 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29627782 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29627775 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P99625 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P09170363 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P09170358 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P09170365 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P09170369 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P09170372 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P09170374 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P09170360 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P09170366 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P09170373 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16686596 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P09170373 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16686597 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P09170372 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16686595 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P99625 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170430 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170384 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170405 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170334 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170398 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170423 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170363 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170435 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170360 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170365 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170366 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170374 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170424 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170396 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170383 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170337 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170395 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16686601 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170369 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16686603 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170335 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170403 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170433 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170411 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170409 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170392 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170427 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170357 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170400 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170407 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170420 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16686606 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09170372 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_203402_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7973583 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7973581 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7973582 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7973580 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 229567_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11727220_at | - | - |
Affymetrix Human Genome U219 Array | 11727220_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1400010582.hg.1 | ENST00000267426.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010582.hg.1 | ENST00000267426.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010582.hg.1 | ENST00000267426.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010582.hg.1 | ENST00000267426.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010582.hg.1 | ENST00000267426.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010582.hg.1 | ENST00000559294.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010582.hg.1 | ENST00000559294.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010582.hg.1 | ENST00000559294.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010582.hg.1 | NM_203402.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010582.hg.1 | NM_203402.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010582.hg.1 | NM_203402.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010582.hg.1 | NM_203402.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400178775.hg.1 | ENST00000267426.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400178775.hg.1 | ENST00000559294.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400178775.hg.1 | NM_203402.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400178775.hg.1 | ENST00000267426.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400178775.hg.1 | ENST00000559294.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400178775.hg.1 | NM_203402.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400178775.hg.1 | TC1400010582.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1400010582.hg.1 | ENST00000267426.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010582.hg.1 | ENST00000267426.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010582.hg.1 | ENST00000267426.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010582.hg.1 | ENST00000267426.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010582.hg.1 | ENST00000267426.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010582.hg.1 | ENST00000559294.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010582.hg.1 | ENST00000559294.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010582.hg.1 | ENST00000559294.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010582.hg.1 | NM_203402.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010582.hg.1 | NM_203402.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010582.hg.1 | NM_203402.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010582.hg.1 | NM_203402.2 | Refseq |
Human Genome U133 Plus 2.0 Array | 229567_at | - | - |
Human Genome U133 Plus 2.0 Array | 229567_PM_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_203402_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229567_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229567_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229567_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229567_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229567_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229567_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229567_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229567_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229567_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229567_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 229567_at5 | - | - |
Human Genome U95 Set | 90117_PM_at | - | - |
Human Genome U95 Set | 90117_at | - | - |
Human Genome U133 Set | 229567_at | - | - |
Human Genome U133 Set | 229567_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051314 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051314 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 205006 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1764557 | BC139911.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1764557 | BQ574746.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1764557 | NM_203402.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1764557 | BC042179.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1764557 | BC139911.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1764557 | BQ574746.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1764557 | NM_203402.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1764557 | BC042179.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1764557 | BC139911.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1764557 | BQ574746.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1764557 | NM_203402.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1764557 | BC042179.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1764557 | BC139911.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1764557 | BQ574746.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1764557 | NM_203402.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1764557 | BC042179.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1764557 | BC139911.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1764557 | BQ574746.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1764557 | NM_203402.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1764557 | BC042179.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_42822881-S | BC139911.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_42822881-S | BQ574746.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_42822881-S | NM_203402.2 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_42822881-S | BC042179.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_6996-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0001990685 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0001990685 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1764557 | BC139911.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1764557 | BQ574746.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1764557 | NM_203402.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1764557 | BC042179.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1764557 | BC139911.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1764557 | BQ574746.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1764557 | NM_203402.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1764557 | BC042179.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 6280193 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_6996 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1764557 | BC139911.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1764557 | BQ574746.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1764557 | NM_203402.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1764557 | BC042179.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1764557 | BC139911.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1764557 | BQ574746.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1764557 | NM_203402.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1764557 | BC042179.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1764557 | BC139911.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1764557 | BQ574746.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1764557 | NM_203402.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1764557 | BC042179.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1764557 | BC139911.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1764557 | BQ574746.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1764557 | NM_203402.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1764557 | BC042179.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1764557 | BC139911.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1764557 | BQ574746.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1764557 | NM_203402.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1764557 | BC042179.1 | human_transcriptome |