Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033152.hg.1 |
TC02002064.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033152.hg.1 |
ENST00000327544.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033152.hg.1 |
ENST00000398146.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033152.hg.1 |
ENST00000420840.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033152.hg.1 |
NM_001024457.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033152.hg.1 |
NM_001078170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033151.hg.1 |
TC02002064.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033151.hg.1 |
ENST00000327544.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033151.hg.1 |
ENST00000398146.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033151.hg.1 |
ENST00000420840.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033151.hg.1 |
NM_001024457.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033151.hg.1 |
NM_001078170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033153.hg.1 |
TC02002064.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033153.hg.1 |
ENST00000327544.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033153.hg.1 |
ENST00000398146.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033153.hg.1 |
ENST00000420840.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033153.hg.1 |
NM_001024457.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033153.hg.1 |
NM_001078170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033154.hg.1 |
TC02002064.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033154.hg.1 |
ENST00000327544.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033154.hg.1 |
ENST00000398146.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033154.hg.1 |
ENST00000420840.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033154.hg.1 |
NM_001024457.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033154.hg.1 |
NM_001078170.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033155.hg.1 |
TC02002064.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033155.hg.1 |
ENST00000327544.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033155.hg.1 |
ENST00000398146.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033155.hg.1 |
ENST00000420840.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033155.hg.1 |
NM_001024457.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033155.hg.1 |
NM_001078170.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200197562.hg.1 |
ENST00000398146.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197562.hg.1 |
NM_001024457.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200197562.hg.1 |
ENST00000398146.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197562.hg.1 |
NM_001024457.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200197562.hg.1 |
TC0200013351.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200197565.hg.1 |
ENST00000398146.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197565.hg.1 |
NM_001024457.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200197565.hg.1 |
ENST00000398146.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197565.hg.1 |
NM_001024457.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200197565.hg.1 |
TC0200013351.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200197564.hg.1 |
ENST00000398146.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197564.hg.1 |
NM_001024457.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200197564.hg.1 |
ENST00000398146.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197564.hg.1 |
NM_001024457.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200197564.hg.1 |
TC0200013351.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200197563.hg.1 |
ENST00000398146.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197563.hg.1 |
NM_001024457.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200197563.hg.1 |
ENST00000398146.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200197563.hg.1 |
NM_001024457.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200197563.hg.1 |
TC0200013351.hg.1 |
Clariom D TC |