Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X019035.hg.1 |
TC0X001435.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X019035.hg.1 |
ENST00000370532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X019035.hg.1 |
NM_004065.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008596.hg.1 |
ENST00000625883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008596.hg.1 |
ENST00000625883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008596.hg.1 |
ENST00000625883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008596.hg.1 |
ENST00000625883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008596.hg.1 |
ENST00000625883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008596.hg.1 |
ENST00000625883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008596.hg.1 |
ENST00000625883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008596.hg.1 |
ENST00000625883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008596.hg.1 |
ENST00000625883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008596.hg.1 |
ENST00000625883.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X019034.hg.1 |
TC0X001435.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X019034.hg.1 |
ENST00000370532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X019034.hg.1 |
NM_004065.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010982.hg.1 |
ENST00000370532.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010982.hg.1 |
ENST00000370532.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010982.hg.1 |
ENST00000370532.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010982.hg.1 |
ENST00000370532.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010982.hg.1 |
ENST00000370532.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010982.hg.1 |
ENST00000370532.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010982.hg.1 |
ENST00000370532.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010982.hg.1 |
NM_004065.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010982.hg.1 |
NM_004065.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010982.hg.1 |
NM_004065.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010982.hg.1 |
NM_004065.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010982.hg.1 |
NM_004065.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174637.hg.1 |
ENST00000370532.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174637.hg.1 |
NM_004065.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174637.hg.1 |
ENST00000370532.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174637.hg.1 |
NM_004065.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174637.hg.1 |
TC0X00010982.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010982.hg.1 |
ENST00000370532.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010982.hg.1 |
ENST00000370532.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010982.hg.1 |
ENST00000370532.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010982.hg.1 |
ENST00000370532.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010982.hg.1 |
ENST00000370532.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010982.hg.1 |
ENST00000370532.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010982.hg.1 |
ENST00000370532.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010982.hg.1 |
NM_004065.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010982.hg.1 |
NM_004065.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010982.hg.1 |
NM_004065.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010982.hg.1 |
NM_004065.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010982.hg.1 |
NM_004065.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174636.hg.1 |
ENST00000370532.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174636.hg.1 |
NM_004065.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174636.hg.1 |
ENST00000370532.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174636.hg.1 |
NM_004065.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174636.hg.1 |
TC0X00010982.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00008596.hg.1 |
ENST00000625883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008596.hg.1 |
ENST00000625883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008596.hg.1 |
ENST00000625883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008596.hg.1 |
ENST00000625883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008596.hg.1 |
ENST00000625883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008596.hg.1 |
ENST00000625883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008596.hg.1 |
ENST00000625883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008596.hg.1 |
ENST00000625883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008596.hg.1 |
ENST00000625883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008596.hg.1 |
ENST00000625883.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174638.hg.1 |
ENST00000370532.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174638.hg.1 |
NM_004065.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174638.hg.1 |
ENST00000370532.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00174638.hg.1 |
NM_004065.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00174638.hg.1 |
TC0X00010982.hg.1 |
Clariom D TC |