Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301829.1 | NP_001288758.1 | 30 | 5523 | 156 | |
NM_005845.4 | NP_005836.2 | 31 | 5664 | 156 | ||
XM_005254025.2 | XP_005254082.1 | 32 | 156 | |||
XM_017020319.1 | XP_016875808.1 | 32 | 156 | |||
XM_017020321.1 | XP_016875810.1 | 21 | 156 | |||
GenBank mRNA | AF339763.1 | - | 1 | 1629 | 156 | |
AY081219.1 | - | 31 | 5552 | 156 | ||
BU689822.1 | - | 1 | 156 | |||
BX647118.1 | - | 21 | 4038 | 156 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |