Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000201586.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000201586.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000201586.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000201586.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000201586.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000201586.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000201586.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000201586.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000201586.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000201586.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000323090.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000323090.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000323090.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000323090.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000323090.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000323090.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000323090.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000323090.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000323090.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000323090.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000594274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000594274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000594274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000594274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000594274.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000597923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000597923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000597923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
ENST00000597923.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
NM_004605.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
NM_004605.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
NM_004605.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
NM_004605.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
NM_004605.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
NM_004605.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
NM_004605.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
NM_004605.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
NM_004605.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
NM_177973.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
NM_177973.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
NM_177973.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
NM_177973.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
NM_177973.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
NM_177973.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
NM_177973.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
NM_177973.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008476.hg.1 |
NM_177973.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009761.hg.1 |
ENST00000201586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009761.hg.1 |
ENST00000323090.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009761.hg.1 |
NM_004605.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009761.hg.1 |
NM_177973.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19009761.hg.1 |
TC19000705.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000201586.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000201586.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000201586.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000201586.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000201586.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000201586.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000201586.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000201586.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000201586.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000201586.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000323090.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000323090.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000323090.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000323090.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000323090.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000323090.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000323090.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000323090.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000323090.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000323090.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000594274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000594274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000594274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000594274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000594274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000597923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000597923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000597923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
ENST00000597923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
NM_004605.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
NM_004605.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
NM_004605.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
NM_004605.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
NM_004605.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
NM_004605.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
NM_004605.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
NM_004605.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
NM_004605.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
NM_177973.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
NM_177973.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
NM_177973.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
NM_177973.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
NM_177973.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
NM_177973.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
NM_177973.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
NM_177973.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008476.hg.1 |
NM_177973.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165263.hg.1 |
ENST00000201586.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165263.hg.1 |
ENST00000323090.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165263.hg.1 |
ENST00000594274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165263.hg.1 |
ENST00000597923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165263.hg.1 |
NM_004605.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165263.hg.1 |
NM_177973.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165263.hg.1 |
TC1900008476.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900165263.hg.1 |
ENST00000201586.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165263.hg.1 |
ENST00000323090.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165263.hg.1 |
ENST00000594274.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165263.hg.1 |
ENST00000597923.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900165263.hg.1 |
NM_004605.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900165263.hg.1 |
NM_177973.1 |
Refseq |