Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
TC01003423.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
ENST00000367979.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
ENST00000367980.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
ENST00000367981.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
ENST00000367982.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
ENST00000367983.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
ENST00000367984.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
ENST00000367985.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
ENST00000412844.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
ENST00000428574.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
ENST00000437437.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
ENST00000442691.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
ENST00000504010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
ENST00000505005.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
ENST00000506209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
ENST00000508740.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
ENST00000511676.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
ENST00000512372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
ENST00000515452.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
ENST00000515621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
NM_001077469.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
NM_001077470.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
NM_001077471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
NM_001077472.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
NM_001077473.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
NM_001077474.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
NM_001077475.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
NM_001077476.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
NM_001077477.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
NM_001077478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
NM_001077479.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
NM_001077480.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
NM_001077481.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
NM_001077482.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01053671.hg.1 |
NM_005122.4 |
Refseq |