Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000315377.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000315377.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000315377.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000315377.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000315377.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000315377.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000315377.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000380381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000380381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000380381.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000380384.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000380384.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000380384.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000380384.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000380384.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000380384.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000380384.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000380384.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000380394.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000380394.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000380394.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000380394.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000380394.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000380394.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000380394.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000498815.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000498815.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
ENST00000498815.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
NM_001010.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
NM_001010.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
NM_001010.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
NM_001010.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
NM_001010.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009654.hg.1 |
NM_001010.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010627.hg.1 |
ENST00000422735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010627.hg.1 |
ENST00000422735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010627.hg.1 |
ENST00000422735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010627.hg.1 |
ENST00000422735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010627.hg.1 |
ENST00000422735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010627.hg.1 |
ENST00000422735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010627.hg.1 |
ENST00000422735.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012541.hg.1 |
TC09000938.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012541.hg.1 |
ENST00000315377.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012541.hg.1 |
ENST00000380384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012541.hg.1 |
ENST00000380394.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012541.hg.1 |
ENST00000498815.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09012541.hg.1 |
NM_001010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010627.hg.1 |
ENST00000422735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010627.hg.1 |
ENST00000422735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010627.hg.1 |
ENST00000422735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010627.hg.1 |
ENST00000422735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010627.hg.1 |
ENST00000422735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010627.hg.1 |
ENST00000422735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010627.hg.1 |
ENST00000422735.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000315377.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000315377.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000315377.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000315377.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000315377.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000315377.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000315377.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000380381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000380381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000380381.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000380384.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000380384.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000380384.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000380384.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000380384.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000380384.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000380384.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000380384.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000380394.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000380394.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000380394.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000380394.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000380394.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000380394.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000380394.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000498815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000498815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
ENST00000498815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
NM_001010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
NM_001010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
NM_001010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
NM_001010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
NM_001010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900009654.hg.1 |
NM_001010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166115.hg.1 |
ENST00000315377.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166115.hg.1 |
ENST00000380384.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166115.hg.1 |
ENST00000380394.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166115.hg.1 |
ENST00000498815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166115.hg.1 |
NM_001010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166115.hg.1 |
ENST00000315377.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166115.hg.1 |
ENST00000380384.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166115.hg.1 |
ENST00000380394.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166115.hg.1 |
ENST00000498815.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900166115.hg.1 |
NM_001010.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900166115.hg.1 |
TC0900009654.hg.1 |
Clariom D TC |