Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000222597.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000222597.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000222597.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000222597.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000222597.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000222597.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000222597.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000222597.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000222597.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000222597.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000415884.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000415884.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000415884.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000415884.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000415884.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000420796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000420796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000420796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000420796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000420796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000420796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000420796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000420796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000420796.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000432748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000432748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000432748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000432748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000432748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000432748.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000479443.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000479443.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000479443.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000479443.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000487517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000487517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000487517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000487517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000487517.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000493361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000493361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000493361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
ENST00000493361.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000222597.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000222597.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000222597.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000222597.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000222597.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000222597.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000222597.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000222597.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000222597.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000222597.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000415884.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000415884.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000415884.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000415884.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000415884.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000420796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000420796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000420796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000420796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000420796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000420796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000420796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000420796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000420796.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000432748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000432748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000432748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000432748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000432748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000432748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000440859.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000479443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000479443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000479443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000479443.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000487517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000487517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000487517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000487517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000487517.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000493361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000493361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000493361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
ENST00000493361.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_001284291.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NM_024814.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008762.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700161970.hg.1 |
ENST00000432748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700161970.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700161970.hg.1 |
ENST00000432748.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700161970.hg.1 |
NR_024199.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700161970.hg.1 |
TC0700008762.hg.1 |
Clariom D TC |