Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF287 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AL359578.1 | - | 1 | 349 | 81 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | leucine rich repeat containing 75A | LRRC75A |
C17orf76, FAM211A |
-73.2kb | Sense | Chr. 17 - 16442005 - 16492191 on NCBI Build GRCh38 |
Downstream | zinc finger protein 624 | ZNF624 | 33.1kb | Sense | Chr. 17 - 16620734 - 16655210 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |