Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC022277.1 | - | 1 | 749 | 102 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | calcium voltage-gated channel auxiliary subunit beta 4 | CACNB4 |
CAB4, CACNLB4, EA5, EIG9, EJM, EJM4, EJM6 |
-74.2kb | Sense | Chr. 2 - 151832771 - 152099988 on NCBI Build GRCh38 |
Downstream | formin like 2 | FMNL2 | FHOD2 | 159.4kb | Anti-sense | Chr. 2 - 152335170 - 152649834 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |