Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001153635.1 | NP_001147107.1 | 3 - 4 | 191 | 117 | |
NM_001153663.1 | NP_001147135.1 | 2 - 3 | 284 | 117 | ||
GenBank mRNA | BC062684.1 | - | 3 - 4 | 191 | 4691440 | 117 |
DB465370.1 | - | 2 - 3 | 233 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |