Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005262220.2 | XP_005262277.1 | 2 | 6038 | 107 | |
XM_011531061.1 | XP_011529363.1 | 3 | 6064 | 107 | ||
GenBank mRNA | AL832136.1 | - | 1 | 4453 | 107 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | collagen type IV alpha 5 chain | COL4A5 |
ASLN, ATS, CA54 |
-22.4kb | Anti-sense | Chr. X - 108439844 - 108697545 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC101928358 | LOC101928358 | 11.6kb | Anti-sense | Chr. X - 108736540 - 108738903 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |