Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
neuroligin 4, Y-linked |
NLGN4Y | -2.0mb | Sense | Chr. Y - 14522578 - 14844945 on NCBI Build GRCh38 | |
Downstream |
family with sequence similarity 41 member A, Y-linked 1 |
FAM41AY1 | FAM41AY | 630.3kb | Sense | Chr. Y - 17500958 - 17516742 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |