Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC023609.2 | - | 1 | 1049 | 73 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | opsin 4 | OPN4 | MOP | -18.8kb | Sense | Chr. 10 - 86653495 - 86666460 on NCBI Build GRCh38 |
Downstream | bone morphogenetic protein receptor type 1A | BMPR1A |
10q23del, ACVRLK3, ALK3, CD292, SKR5 |
69.1kb | Sense | Chr. 10 - 86755786 - 86927969 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |