Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
ENST00000295834.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
ENST00000295834.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
ENST00000295834.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
ENST00000295834.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
ENST00000295834.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
ENST00000295834.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
ENST00000295834.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
ENST00000393750.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
ENST00000393750.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
ENST00000393750.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
ENST00000393750.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
ENST00000393750.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
ENST00000393750.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
ENST00000472846.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
ENST00000472846.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
ENST00000472846.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
ENST00000495375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
ENST00000495375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
ENST00000495375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
ENST00000495375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
ENST00000495375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
ENST00000495375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
ENST00000495375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
NM_001443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
NM_001443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
NM_001443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
NM_001443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
NM_001443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
NM_001443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013362.hg.1 |
NM_001443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
ENST00000295834.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
ENST00000295834.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
ENST00000295834.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
ENST00000295834.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
ENST00000295834.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
ENST00000295834.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
ENST00000295834.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
ENST00000393750.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
ENST00000393750.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
ENST00000393750.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
ENST00000393750.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
ENST00000393750.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
ENST00000393750.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
ENST00000472846.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
ENST00000472846.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
ENST00000472846.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
ENST00000495375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
ENST00000495375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
ENST00000495375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
ENST00000495375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
ENST00000495375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
ENST00000495375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
ENST00000495375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
NM_001443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
NM_001443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
NM_001443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
NM_001443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
NM_001443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
NM_001443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013362.hg.1 |
NM_001443.2 |
Refseq |