Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cytochrome c oxidase assembly factor 1 homolog | COA1 |
C7orf44, MITRAC15 |
-29.8kb | Anti-sense | Chr. 7 - 43608452 - 43729541 on NCBI Build GRCh38 |
Downstream | mitochondrial ribosomal protein S24 | MRPS24 |
HSPC335, MRP-S24, S24mt, bMRP-47, bMRP47 |
100.6kb | Anti-sense | Chr. 7 - 43866558 - 43869546 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Homo sapiens Probe spans exons |