Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | IQ motif containing GTPase activating protein 2 | IQGAP2 | Sense | Chr. 5 - 76403255 - 76708132 on NCBI Build GRCh38 | ||
Downstream |
non-protein coding RNA, upstream of F2R/PAR1 |
NCRUPAR |
NCRNA00193, ncR-uPAR |
23.1kb | Sense | Chr. 5 - 76712403 - 76712888 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |