Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NETO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138966.3 | NP_620416.1 | 11 | 5953 | 110 | |
XM_005266774.4 | XP_005266831.1 | 11 | 5472 | 110 | ||
XM_017026020.1 | XP_016881509.1 | 11 | 5560 | 110 | ||
GenBank mRNA | AK123319.1 | - | 1 | 1850 | 110 | |
BU739516.1 | - | 1 | 110 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cerebellin 2 precursor | CBLN2 | -105.3kb | Sense | Chr. 18 - 72533720 - 72638485 on NCBI Build GRCh38 | |
Downstream | microRNA 548av | MIR548AV | 107.4kb | Sense | Chr. 18 - 72853321 - 72853382 on NCBI Build GRCh38 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon |