Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RNA binding protein with serine rich domain 1 | RNPS1 | E5.1 | Sense | Chr. 16 - 2253116 - 2268412 on NCBI Build GRCh38 | |
Downstream | microRNA 3677 | MIR3677 | mir-3677 | 12.0kb | Anti-sense | Chr. 16 - 2270713 - 2270772 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |