Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transmembrane protein 245 | TMEM245 |
C9orf5, CG-2, CG2 |
-12.8kb | Sense | Chr. 9 - 109015135 - 109119963 on NCBI Build GRCh38 |
Downstream | ferric chelate reductase 1 like | FRRS1L |
C9orf4, CG-6, CG6, EIEE37 |
1.0kb | Sense | Chr. 9 - 109136188 - 109167291 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |