Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | C-type lectin domain family 16 member A | CLEC16A |
Gop-1, KIAA0350 |
-14.5kb | Sense | Chr. 16 - 10944488 - 11193272 on NCBI Build GRCh38 |
Downstream | suppressor of cytokine signaling 1 | SOCS1 |
CIS1, CISH1, JAB, SOCS-1, SSI-1, SSI1, TIP3 |
45.4kb | Anti-sense | Chr. 16 - 11254417 - 11256182 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |