Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | family with sequence similarity 228 member A | FAM228A | C2orf84 | -91.5kb | Anti-sense | Chr. 2 - 24175043 - 24191698 on NCBI Build GRCh38 |
Downstream | nuclear receptor coactivator 1 | NCOA1 |
F-SRC-1, KAT13A, RIP160, SRC1, bHLHe42, bHLHe74 |
206.6kb | Anti-sense | Chr. 2 - 24492050 - 24770702 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |