Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | anthrax toxin receptor-like pseudogene 1 | ANTXRLP1 | Sense | Chr. 10 - 46219025 - 46269608 on NCBI Build GRCh38 | ||
Downstream | basic proline-rich protein-like | LOC105378577 | 35.0kb | Anti-sense | Chr. 10 - 46283571 - 46285997 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |