Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | HOXA10-HOXA9 readthrough | HOXA10-HOXA9 | -4.8kb | Anti-sense | Chr. 7 - 27162438 - 27180261 on NCBI Build GRCh38 | |
Downstream | homeobox A13 | HOXA13 |
HOX1, HOX1J |
7.7kb | Anti-sense | Chr. 7 - 27196880 - 27200106 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |