Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | translation machinery associated 16 homolog | TMA16 | C4orf43 | -149.1kb | Anti-sense | Chr. 4 - 163494521 - 163520539 on NCBI Build GRCh38 |
Downstream | acidic nuclear phosphoprotein 32 family member C | ANP32C | PP32R1 | 525.8kb | Sense | Chr. 4 - 164197007 - 164197711 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |