Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | neutral cholesterol ester hydrolase 1 | NCEH1 |
AADACL1, NCEH |
Sense | Chr. 3 - 172630645 - 172711218 on NCBI Build GRCh38 | |
Downstream | epithelial cell transforming 2 | ECT2 | ARHGEF31 | 54.5kb | Anti-sense | Chr. 3 - 172750682 - 172829273 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |