Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | phosphatase and actin regulator 4 | PHACTR4 | PPP1R124 | Sense | Chr. 1 - 28369574 - 28500369 on NCBI Build GRCh38 | |
Downstream | regulator of chromosome condensation 1 | RCC1 |
CHC1, RCC1-I, SNHG3-RCC1 |
22.1kb | Sense | Chr. 1 - 28505943 - 28539196 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |